Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000745.3 | NP_000736.2 | 5 | 900 | ||
XM_017021881.1 | XP_016877370.1 | 5 | 899 | |||
GenBank mRNA | AF385586.1 | - | 5 | 846 | ||
BC033639.1 | - | 5 | 862 | 5501542 | ||
M83712.1 | - | 5 | 848 | |||
U62434.1 | - | 5 | 854 | |||
Y08419.1 | - | 5 | 700 |