Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000047.2 | NP_000038.2 | 6 | 903 | ||
NM_001282628.1 | NP_001269557.1 | 7 | 1189 | |||
NM_001282631.1 | NP_001269560.1 | 5 | 673 | |||
XM_005274518.2 | XP_005274575.1 | 6 | 830 | |||
XM_005274519.4 | XP_005274576.1 | 6 | 993 | |||
XM_005274521.4 | XP_005274578.1 | 5 | 786 | |||
XM_017029525.1 | XP_016885014.1 | 7 | 1120 | |||
GenBank mRNA | AK223183.1 | - | 6 | 733 | ||
AK223199.1 | - | 6 | 700 | |||
AK293648.1 | - | 5 | 608 | |||
AK301004.1 | - | 7 | 1020 | |||
AK313093.1 | - | 6 | 722 | |||
BC130438.1 | - | 6 | 698 | 40146469 | ||
X83573.1 | - | 6 | 720 |