Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281726.1 | NP_001268655.1 | 2 | 306 | ||
XM_006713314.3 | XP_006713377.1 | 3 | 586 | |||
XM_011534041.2 | XP_011532343.1 | 3 | 423 | |||
XM_017007088.1 | XP_016862577.1 | 4 | 699 | |||
GenBank mRNA | AK294301.1 | - | 4 | 438 | ||
AK297033.1 | - | 2 | 298 | |||
AK301838.1 | - | 2 | 498 | |||
AK313252.1 | - | 2 | 289 | |||
BC012099.1 | - | 2 | 232 | 4559618 | ||
BE814688.1 | - | 2 | 329 | |||
BM919775.1 | - | 2 | 299 | |||
BP326502.1 | - | 2 | 306 | |||
U03274.1 | - | 2 | 235 |