Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000161.2 | NP_000152.1 | 5, 6 | 791 | ||
NM_001024024.1 | NP_001019195.1 | 5, 6 | 791 | |||
XM_017021218.1 | XP_016876707.1 | 5, 6 | 1223 | |||
GenBank mRNA | AF321276.1 | - | 2, 3 | 115 | ||
AY137463.1 | - | 5, 6 | 413 | |||
BC025415.2 | - | 5, 6 | 778 | 4129035 | ||
CR536551.1 | - | 5, 6 | 630 | |||
S44049.1 | - | 5, 6 | 604 | |||
U19523.1 | - | 5, 6 | 778 | |||
U66097.1 | - | 5, 6 | 783 | |||
Z16418.1 | - | 5, 6 | 454 | |||
Z29434.2 | - | 5, 6 | 777 |