Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000169.2 | NP_000160.1 | 6 | 1063 | ||
GenBank mRNA | AK222627.1 | - | 6 | 975 | ||
AK291095.1 | - | 6 | 982 | |||
BC002689.2 | - | 6 | 959 | 3609235 | ||
BT006864.1 | - | 6 | 953 | |||
D00039.1 | - | 6 | 877 | |||
X05790.1 | - | 6 | 976 | |||
X16889.1 | - | 6 | 955 |