Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126131.1 | NP_001119603.1 | 5 | 1352 | ||
NM_002693.2 | NP_002684.1 | 5 | 1364 | |||
GenBank mRNA | AK291281.1 | - | 5 | 1383 | ||
BC042571.1 | - | 5 | 1349 | 4384475 | ||
BC050559.1 | - | 5 | 1333 | 6042595 | ||
D84103.1 | - | 4 | 1188 | |||
U60325.1 | - | 4 | 1241 | |||
X98093.1 | - | 5 | 1364 |