Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDUA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000203.4 | NP_000194.2 | 6 | 825 | ||
NR_110313.1 | 6 | 825 | ||||
XM_011513461.2 | XP_011511763.1 | 5 | 970 | |||
XM_017008163.1 | XP_016863652.1 | 5 | 1279 | |||
GenBank mRNA | AK098805.1 | - | 5 | 793 | ||
AK125223.1 | - | 5 | 637 | |||
AK291816.1 | - | 6 | 758 | |||
BC029959.1 | - | 6 | 749 | |||
M74715.1 | - | 6 | 825 |