Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161.4 | NP_001152.1 | 5 | 964 | ||
NM_001244390.1 | NP_001231319.1 | 3 | 716 | |||
NM_147172.2 | NP_671701.1 | 5 | 893 | |||
NM_147173.2 | NP_671702.1 | 4 | 851 | |||
GenBank mRNA | BC004926.2 | - | 3 | 761 | 3623134 | |
U30313.1 | - | 4 | 780 |