Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281501.1 | NP_001268430.1 | 2, 3 | 334 | ||
NM_001281502.1 | NP_001268431.1 | 2, 3 | 403 | |||
NM_004265.3 | NP_004256.1 | 2, 3 | 675 | |||
GenBank mRNA | AF084559.1 | - | 2, 3 | 467 | ||
AF108658.1 | - | 2, 3 | 260 | |||
AF126799.1 | - | 2, 3 | 330 | |||
AK074939.1 | - | 2, 3 | 407 | |||
AK074991.1 | - | 2, 3 | 143 | |||
AK290291.1 | - | 2, 3 | 281 | |||
AK299762.1 | - | 2, 3 | 403 | |||
BC009011.1 | - | 2, 3 | 409 | 4181037 | ||
HY090295.1 | - | 2, 3 | 335 |