Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004278.3 | NP_004269.1 | 5, 6 | 551 | ||
XM_017025352.1 | XP_016880841.1 | 5, 6 | 537 | |||
XM_017025353.1 | XP_016880842.1 | 5, 6 | 537 | |||
GenBank mRNA | AB017165.1 | - | 5, 6 | 524 | ||
AK292932.1 | - | 5, 6 | 536 | |||
BC068197.1 | - | 5, 6 | 530 | 6711166 |