Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004666.2 | NP_004657.2 | 4 | 568 | ||
GenBank mRNA | AJ132099.1 | - | 4 | 562 | ||
AK290425.1 | - | 4 | 565 | |||
AK314265.1 | - | 4 | 565 | |||
BC096265.3 | - | 4 | 562 | 40006204 | ||
BC096266.3 | - | 4 | 562 | 40006205 | ||
BC096267.3 | - | 4 | 562 | 40006207 | ||
BC096268.1 | - | 4 | 561 | 40006208 | ||
BJ995136.1 | - | 4 | 568 | |||
U39664.1 | - | 4 | 563 |