Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100397.1 | NP_001093867.1 | 3, 4 | 485 | ||
NM_001282899.1 | NP_001269828.1 | 3, 4 | 356 | |||
NM_007023.3 | NP_008954.2 | 6, 7 | 726 | |||
XM_017003196.1 | XP_016858685.1 | 6, 7 | 593 | |||
GenBank mRNA | AB027471.1 | - | 6, 7 | 596 | ||
AB209681.1 | - | 6, 7 | 655 | |||
AK294445.1 | - | 3, 4 | 205 | |||
AK294997.1 | - | 6, 7 | 691 | |||
AK304278.1 | - | 3, 4 | 245 | |||
AK311500.1 | - | 3, 4 | 485 | |||
AK311697.1 | - | 3, 4 | 462 | |||
AK313084.1 | - | 6, 7 | 595 | |||
AK316014.1 | - | 3, 4 | 320 | |||
AK316072.1 | - | 3, 4 | 252 | |||
BC024004.2 | - | 6, 7 | 565 | 4823935 | ||
BC040183.2 | - | 3, 4 | 486 | 4801092 | ||
DC335305.1 | - | 3, 4 | 356 | |||
KJ534924.1 | - | 6, 7 | 503 | |||
KJ534978.1 | - | 6, 7 | 539 | |||
U78516.1 | - | 6, 7 | 647 |