Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012294.3 | NP_036426.3 | 11 | 1020 | ||
XM_005249914.2 | XP_005249971.1 | 11 | 1239 | |||
XM_005249916.1 | XP_005249973.1 | 11 | 1183 | |||
XM_011515652.2 | XP_011513954.1 | 3 | 2993 | |||
XM_017012837.1 | XP_016868326.1 | 9 | 924 | |||
GenBank mRNA | AK295859.1 | - | 1 | 505 | ||
AL833195.1 | - | 11 | 1321 | |||
BC039203.1 | - | 1 | 331 | 4811483 | ||
BP349215.1 | - | 2 | 78 | |||
D87467.1 | - | 1 | 312 | |||
KU178490.1 | - | 1 | 257 | |||
KU178491.1 | - | 1 | 257 |