Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130955.1 | NP_001124427.1 | 9 | 1887 | ||
NM_015318.3 | NP_056133.2 | 10 | 1366 | |||
XM_005272464.4 | XP_005272521.1 | 19 | 2517 | |||
XM_006722705.3 | XP_006722768.1 | 19 | 2649 | |||
XM_006722706.3 | XP_006722769.1 | 20 | 2812 | |||
XM_006722708.2 | XP_006722771.1 | 10 | 1494 | |||
XM_011527835.2 | XP_011526137.1 | 19 | 2517 | |||
XM_011527836.2 | XP_011526138.1 | 19 | 2517 | |||
XM_011527837.2 | XP_011526139.1 | 19 | 2517 | |||
XM_011527838.2 | XP_011526140.1 | 18 | 2205 | |||
XM_011527839.2 | XP_011526141.1 | 18 | 2271 | |||
XM_011527840.1 | XP_011526142.1 | 10 | 1366 | |||
XM_011527841.2 | XP_011526143.1 | 19 | 2510 | |||
GenBank mRNA | AB011093.1 | - | 10 | 1267 | ||
AK074372.1 | - | 5 | 567 | |||
AK127045.1 | - | 9 | 1887 | |||
BC077721.1 | - | 11 | 1434 | 5921159 |