Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015320.3 | NP_056135.2 | 10 | 1874 | ||
NM_032995.2 | NP_127462.1 | 9 | 1463 | |||
XM_005263681.1 | XP_005263738.1 | 10 | 4951 | |||
XM_005263687.3 | XP_005263744.1 | 7 | 1165 | |||
XM_005263688.2 | XP_005263745.1 | 6 | 1095 | |||
XM_005263689.4 | XP_005263746.1 | 7 | 1010 | |||
XM_011511274.2 | XP_011509576.1 | 10 | 4951 | |||
XM_011511276.2 | XP_011509578.1 | 6 | 813 | |||
XM_011511277.1 | XP_011509579.1 | 6 | 940 | |||
GenBank mRNA | AB029035.1 | - | 9 | 1429 | ||
AB042199.1 | - | 7 | 1154 | |||
AF249745.1 | - | 10 | 1875 | |||
AL137289.1 | - | 5 | 775 | |||
DQ424905.1 | - | 7 | 1175 |