Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303533.1 | NP_001290462.1 | 6 | 638 | ||
NM_001303534.1 | NP_001290463.1 | 6 | 733 | |||
NM_016339.4 | NP_057423.1 | 6 | 918 | |||
XM_006721932.3 | XP_006721995.1 | 6 | 1412 | |||
XM_006721935.3 | XP_006721998.1 | 7 | 689 | |||
XM_006721937.3 | XP_006722000.1 | 6 | 2514 | |||
XM_006721938.3 | XP_006722001.1 | 3 | 305 | |||
GenBank mRNA | AF117946.1 | - | 6 | 553 | ||
AK294642.1 | - | 2 | 540 | |||
AK304160.1 | - | 6 | 733 | |||
AK313492.1 | - | 6 | 593 | |||
BC029956.1 | - | 6 | 1412 | |||
BC115374.1 | - | 4 | 447 | 40028278 | ||
BC115375.1 | - | 4 | 447 | 40028280 |