Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA5L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323640.1 | NP_001310569.1 | 4 | 1679 | ||
NM_024063.2 | NP_076968.2 | 4 | 1679 | |||
NR_027635.1 | 4 | 1679 | ||||
NR_136645.1 | 4 | 1686 | ||||
NR_136646.1 | 5 | 1874 | ||||
NR_136647.1 | 5 | 1841 | ||||
NR_136648.1 | 5 | 1874 | ||||
GenBank mRNA | AK023232.1 | - | 4 | 1625 | ||
AK225463.1 | - | 4 | 1625 | |||
AK291457.1 | - | 4 | 1632 | |||
BC000981.2 | - | 4 | 1612 | 2985725 | ||
BC051861.2 | - | 4 | 1624 |