Search Thermo Fisher Scientific
Gene Symbol: DDAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303007.1 | NP_001289936.1 | 6 | 1865 | ||
NM_001303008.1 | NP_001289937.1 | 7 | 965 | |||
NM_013974.2 | NP_039268.1 | 7 | 1040 | |||
XM_011514448.2 | XP_011512750.1 | 7 | 1116 | |||
GenBank mRNA | AF070667.1 | - | 7 | 1037 | ||
AF087894.1 | - | 7 | 1037 | |||
AK026191.1 | - | 6 | 1392 | |||
AK098671.1 | - | 7 | 1027 | |||
BC001435.2 | - | 7 | 952 | 3138832 | ||
FJ224290.1 | - | 7 | 1037 |