Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290137.1 | NP_001277066.1 | 9 | 1167 | ||
NM_001290139.1 | NP_001277068.1 | 9 | 1237 | |||
NM_001323635.1 | NP_001310564.1 | 9 | 1272 | |||
NM_001323636.1 | NP_001310565.1 | 8 | 1192 | |||
NM_032304.3 | NP_115680.1 | 8 | 1198 | |||
XM_005255632.1 | XP_005255689.1 | 8 | 1405 | |||
XM_011522712.1 | XP_011521014.1 | 8 | 1429 | |||
XM_017023774.1 | XP_016879263.1 | 8 | 1261 | |||
GenBank mRNA | AK054841.1 | - | 6 | 1493 | ||
AK098340.1 | - | 3 | 2025 | |||
BC004353.1 | - | 8 | 1173 | 3351990 | ||
BC015008.1 | - | 8 | 1126 | 4862387 | ||
BC033796.1 | - | 7 | 1527 | 5173915 | ||
BC036747.1 | - | 7 | 1527 | |||
BI834617.1 | - | 4 | 506 | |||
BQ067068.1 | - | 4 | 485 |