Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000152.4 | NP_000143.2 | 20 | 3460 | ||
NM_001079803.2 | NP_001073271.1 | 21 | 3275 | |||
NM_001079804.2 | NP_001073272.1 | 20 | 3195 | |||
XM_005257193.2 | XP_005257250.1 | 21 | 3261 | |||
XM_005257194.4 | XP_005257251.1 | 21 | 3301 | |||
GenBank mRNA | BC040431.1 | - | 21 | 3207 | 5739991 | |
M34424.1 | - | 19 | 3095 | |||
Y00839.1 | - | 20 | 3282 |