Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000157.3 | NP_000148.2 | 9 | 1462 | ||
NM_001005741.2 | NP_001005741.1 | 10 | 1721 | |||
NM_001005742.2 | NP_001005742.1 | 10 | 1702 | |||
NM_001171811.1 | NP_001165282.1 | 8 | 1538 | |||
NM_001171812.1 | NP_001165283.1 | 8 | 1312 | |||
GenBank mRNA | AK291911.1 | - | 10 | 1700 | ||
AK298900.1 | - | 8 | 1312 | |||
AK300186.1 | - | 8 | 1382 | |||
AK300829.1 | - | 8 | 1375 | |||
AK300876.1 | - | 10 | 1702 | |||
AK301374.1 | - | 7 | 1071 | |||
AK302000.1 | - | 8 | 1287 | |||
AK312502.1 | - | 10 | 1518 | |||
BC003356.1 | - | 9 | 1401 | 2899915 | ||
BX648487.1 | - | 10 | 1530 | |||
D13286.1 | - | 9 | 1401 | |||
K02920.1 | - | 8 | 1281 | |||
KJ690771.1 | - | 9 | 1345 | |||
M16328.1 | - | 9 | 1731 | |||
M19285.1 | - | 9 | 1418 |
Gene Symbol: GBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KJ690774.1 | - | 1096 | |||
KJ690776.1 | - | 986 |