Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000387.5 | NP_000378.1 | 5 | 647 | ||
GenBank mRNA | AK301744.1 | - | 6 | 608 | ||
AK312962.1 | - | 5 | 540 | |||
BC001689.2 | - | 5 | 519 | 3050073 | ||
BI913588.1 | - | 5 | 550 | |||
Y10319.1 | - | 5 | 510 |