Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256414.1 | NP_001243343.1 | 6 | 857 | ||
NM_002069.5 | NP_002060.4 | 6 | 994 | |||
GenBank mRNA | AF055013.1 | - | 6 | 669 | ||
AF493905.1 | - | 6 | 657 | |||
AK292953.1 | - | 6 | 968 | |||
AK304442.1 | - | 6 | 857 | |||
AL049933.1 | - | 6 | 972 | |||
BT019775.1 | - | 6 | 657 | |||
M17219.1 | - | 6 | 642 |