Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002073.2 | NP_002064.1 | 3 | 2257 | ||
GenBank mRNA | BC026342.1 | - | 3 | 2257 | 4814858 | |
BC037333.1 | - | 3 | 2395 | 5264642 | ||
BC078163.1 | - | 2 | 2095 | 6180193 | ||
J03260.1 | - | 2 | 1729 |