Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC35C2 SIRNA Assays ›
Gene Symbol:
SLC35C2
Gene Name:
solute carrier family 35 member C2
Gene Aliases:
BA394O2.1, C20orf5, CGI-15, OVCOV1
Chromosome Location:
Chr.20: 46349528 - 46364461 on Build GRCh38
UniGene ID:
Hs.593344
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119784
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC35C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281457.1
NP_001268386.1
10
1511
NM_001281458.1
NP_001268387.1
11
1450
NM_001281459.1
NP_001268388.1
10
1448
NM_001281460.1
NP_001268389.1
11
1914
NM_015945.11
NP_057029.8
10
1582
NM_173073.3
NP_775096.1
10
1519
NM_173179.3
NP_775271.1
10
1777
XM_011528831.1
XP_011527133.1
10
1760
XM_011528832.1
XP_011527134.1
10
2054
XM_011528833.1
XP_011527135.1
10
1578
XM_011528834.1
XP_011527136.1
10
1579
XM_011528835.1
XP_011527137.1
9
1468
XM_011528836.1
XP_011527138.1
10
1663
XM_011528837.1
XP_011527139.1
10
1497
XM_011528838.1
XP_011527140.1
8
1616
XM_017027856.1
XP_016883345.1
10
2477
XM_017027857.1
XP_016883346.1
10
2005
XM_017027858.1
XP_016883347.1
10
1915
XM_017027859.1
XP_016883348.1
10
2282
XM_017027860.1
XP_016883349.1
11
1722
XM_017027861.1
XP_016883350.1
10
1681
XM_017027862.1
XP_016883351.1
9
1856
GenBank mRNA
AF132949.1
-
10
1518
AF455052.1
-
10
1537
AK094358.1
-
2
1780
AK128842.1
-
2
2786
AK222928.1
-
10
1522
AK300207.1
-
10
1465
AK300798.1
-
10
1380
AK301022.1
-
11
1392
BC014191.2
-
10
1463
4761663
BC021138.1
-
10
1720
4870715
BC025277.1
-
10
1778
4865888
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
negative regulation of gene expression
UDP-glucose transport
protein O-linked fucosylation
positive regulation of Notch signaling pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0