Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GANAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278192.1 | NP_001265121.1 | 2 | 279 | ||
NM_001278193.1 | NP_001265122.1 | 2 | 279 | |||
NM_001278194.1 | NP_001265123.1 | 4 | 512 | |||
NM_198334.2 | NP_938148.1 | 5 | 621 | |||
NM_198335.3 | NP_938149.2 | 5 | 621 | |||
XM_017017412.1 | XP_016872901.1 | 3 | 355 | |||
XM_017017413.1 | XP_016872902.1 | 5 | 684 | |||
XM_017017414.1 | XP_016872903.1 | 5 | 515 | |||
GenBank mRNA | AF144074.1 | - | 5 | 510 | ||
AJ000332.1 | - | 5 | 494 | |||
AK026997.1 | - | 5 | 511 | |||
AK295806.1 | - | 2 | 167 | |||
AK299819.1 | - | 4 | 395 | |||
AK302752.1 | - | 2 | 185 | |||
AK314370.1 | - | 5 | 510 | |||
BC065266.1 | - | 4 | 388 | 6155944 | ||
BG720650.1 | - | 5 | 566 | |||
D42041.1 | - | 5 | 492 | |||
EU794613.1 | - | 5 | 527 |