Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000157.3 | NP_000148.2 | 11 | 1689 | ||
NM_001005741.2 | NP_001005741.1 | 12 | 1948 | |||
NM_001005742.2 | NP_001005742.1 | 12 | 1929 | |||
NM_001171811.1 | NP_001165282.1 | 10 | 1765 | |||
NM_001171812.1 | NP_001165283.1 | 10 | 1539 | |||
GenBank mRNA | AK291911.1 | - | 12 | 1927 | ||
AK298900.1 | - | 10 | 1539 | |||
AK300186.1 | - | 10 | 1609 | |||
AK300829.1 | - | 10 | 1602 | |||
AK300876.1 | - | 12 | 1929 | |||
AK301374.1 | - | 9 | 1298 | |||
AK302000.1 | - | 10 | 1514 | |||
AK312502.1 | - | 12 | 1745 | |||
BC003356.1 | - | 11 | 1628 | 2899915 | ||
BX648487.1 | - | 12 | 1757 | |||
D13286.1 | - | 11 | 1628 | |||
K02920.1 | - | 10 | 1508 | |||
KJ690771.1 | - | 11 | 1572 | |||
KJ690772.1 | - | 12 | 1407 | |||
KJ690773.1 | - | 11 | 1517 | |||
M16328.1 | - | 11 | 1958 | |||
M19285.1 | - | 11 | 1645 |
Gene Symbol: GBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002188.3 | 13 | 1405 | |||
GenBank mRNA | AK295259.1 | - | 10 | 1350 | ||
AK297653.1 | - | 10 | 1325 | |||
BC000349.2 | - | 11 | 1242 | |||
BC030240.1 | - | 13 | 1284 | |||
BC092482.1 | - | 11 | 1240 | |||
KJ690774.1 | - | 9 | 1323 | |||
KJ690775.1 | - | 11 | 1158 | |||
KJ690776.1 | - | 10 | 1213 |