Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC28A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004212.3 | NP_004203.2 | 12 | 1207 | ||
XM_011522198.2 | XP_011520500.1 | 12 | 1227 | |||
XM_011522199.1 | XP_011520501.1 | 12 | 1178 | |||
XM_011522200.2 | XP_011520502.1 | 12 | 1178 | |||
XM_011522201.2 | XP_011520503.1 | 12 | 1178 | |||
XM_011522202.2 | XP_011520504.1 | 4 | 378 | |||
GenBank mRNA | AF036109.1 | - | 11 | 1156 | ||
AK222709.1 | - | 12 | 1207 | |||
AK291974.1 | - | 12 | 1178 | |||
BC093737.1 | - | 11 | 1170 | 7939582 | ||
BC112047.1 | - | 11 | 1182 | 8327515 | ||
U84392.1 | - | 12 | 1201 |