Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002703.4 | NP_002694.3 | 2, 3 | 362 | ||
GenBank mRNA | AK222759.1 | - | 2, 3 | 301 | ||
AK290942.1 | - | 2, 3 | 217 | |||
AK312297.1 | - | 2, 3 | 338 | |||
BC004200.2 | - | 2, 3 | 234 | 3533162 | ||
D13757.1 | - | 2, 3 | 324 | |||
DB039655.1 | - | 2, 3 | 362 | |||
U00238.1 | - | 2, 3 | 253 |