Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307985.1 | NP_001294914.1 | 8 | 2069 | ||
NM_018484.3 | NP_060954.1 | 10 | 2393 | |||
XM_011545167.1 | XP_011543469.1 | 9 | 2043 | |||
GenBank mRNA | AB026116.1 | - | 10 | 2046 | ||
AK075224.1 | - | 10 | 1739 | |||
AK129930.1 | - | 8 | 1981 | |||
AK290791.1 | - | 10 | 2376 | |||
BC034384.1 | - | 10 | 2368 | 5190509 |