Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174089.1 | NP_001167560.1 | 4, 5 | 393 | ||
NM_001174090.1 | NP_001167561.1 | 4, 5 | 543 | |||
NM_032034.3 | NP_114423.1 | 3, 4 | 385 | |||
NR_135000.1 | 4, 5 | 457 | ||||
XM_005260856.4 | XP_005260913.1 | 4, 5 | 881 | |||
XM_005260857.1 | XP_005260914.1 | 4, 5 | 412 | |||
XM_011529383.2 | XP_011527685.1 | 4, 5 | 1988 | |||
XM_011529384.1 | XP_011527686.1 | 4, 5 | 630 | |||
XM_011529385.1 | XP_011527687.1 | 4, 5 | 457 | |||
XM_017028093.1 | XP_016883582.1 | 4, 5 | 881 | |||
XM_017028094.1 | XP_016883583.1 | 4, 5 | 265 | |||
XM_017028095.1 | XP_016883584.1 | 4, 5 | 736 | |||
XM_017028096.1 | XP_016883585.1 | 5, 6 | 642 | |||
XM_017028097.1 | XP_016883586.1 | 4, 5 | 881 | |||
GenBank mRNA | AF336127.1 | - | 3, 4 | 385 | ||
AK091157.1 | - | 4, 5 | 457 | |||
AK296508.1 | - | 4, 5 | 543 | |||
AK296760.1 | - | 4, 5 | 342 | |||
BC110540.1 | - | 3, 4 | 358 | 40035522 | ||
BC110541.1 | - | 3, 4 | 358 | |||
DA630556.1 | - | 4, 5 | 457 | |||
DC341640.1 | - | 4, 5 | 393 |