Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 1411 | |||
NM_001318156.1 | NP_001305085.1 | 1234 | ||||
NM_001318157.1 | NP_001305086.1 | 2087 | ||||
NM_001318158.1 | NP_001305087.1 | 1769 | ||||
NR_134513.1 | 1526 | |||||
NR_134514.1 | 1645 | |||||
NR_134515.1 | 1796 | |||||
GenBank mRNA | AB006190.1 | - | 1248 | |||
DQ786216.1 | - | 623 |