Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174166.1 | NP_001167637.1 | 5 | 793 | ||
NM_004694.4 | NP_004685.2 | 4 | 666 | |||
XM_005257789.3 | XP_005257846.2 | 4 | 831 | |||
XM_011525461.2 | XP_011523763.1 | 4 | 510 | |||
XM_017025291.1 | XP_016880780.1 | 5 | 630 | |||
XM_017025292.1 | XP_016880781.1 | 4 | 693 | |||
GenBank mRNA | AK290417.1 | - | 4 | 542 | ||
AK313121.1 | - | 4 | 532 | |||
AX750581.1 | - | 5 | 645 | |||
AX750582.1 | - | 3 | 480 | |||
BC064832.1 | - | 4 | 668 | 6498761 | ||
U79745.1 | - | 5 | 645 |