Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001677.3 | NP_001668.1 | 2 | 324 | ||
GenBank mRNA | AK290181.1 | - | 2 | 337 | ||
AK296059.1 | - | 2 | 212 | |||
AK316043.1 | - | 2 | 269 | |||
AY545986.1 | - | 2 | 203 | |||
BC000006.2 | - | 2 | 289 | 3506311 | ||
BQ934151.1 | - | 3 | 528 | |||
BT009787.1 | - | 2 | 203 | |||
CR407616.1 | - | 2 | 203 | |||
U16799.1 | - | 2 | 248 |