Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002811.4 | NP_002802.2 | 1, 2 | 220 | ||
GenBank mRNA | AK315381.1 | - | 1, 2 | 178 | ||
BC000338.2 | - | 1, 2 | 207 | |||
BC012606.1 | - | 1, 2 | 201 | 4334058 | ||
BU601564.1 | - | 1, 2 | 223 |