Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003348.3 | NP_003339.1 | 4 | 1751 | ||
GenBank mRNA | AK098233.1 | - | 1 | 2187 | ||
AL110140.1 | - | 1 | 62 |