Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195328.1 | NP_001182257.1 | 2 | 307 | ||
NM_004251.4 | NP_004242.1 | 3 | 396 | |||
XM_017029964.1 | XP_016885453.1 | 3 | 798 | |||
GenBank mRNA | AF498944.1 | - | 1 | 113 | ||
AK290505.1 | - | 2 | 286 | |||
BC017265.2 | - | 3 | 295 | 4139714 | ||
CR450362.1 | - | 1 | 113 | |||
DA141710.1 | - | 3 | 421 | |||
U44103.1 | - | 1 | 113 |