Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204077.1 | NP_001191006.1 | 4 | 448 | ||
NM_004788.3 | NP_004779.2 | 4 | 448 | |||
GenBank mRNA | AF087968.1 | - | 4 | 387 | ||
AK296369.1 | - | 4 | 395 | |||
AL713515.1 | - | 4 | 447 | |||
BC030973.1 | - | 4 | 397 | |||
BC052643.1 | - | 4 | 406 | |||
BC062695.1 | - | 4 | 428 | |||
BC111417.1 | - | 4 | 422 | 40038776 | ||
BC112367.1 | - | 4 | 425 | 40038767 | ||
BU167020.1 | - | 4 | 405 | |||
D50916.1 | - | 4 | 385 |