Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002800.4 | NP_002791.1 | 4 | 413 | ||
GenBank mRNA | AK303118.1 | - | 3 | 284 | ||
BC065513.1 | - | 4 | 368 | 5749985 | ||
BI832878.1 | - | 4 | 395 | |||
BX641100.1 | - | 2 | 2094 | |||
CR541656.1 | - | 4 | 344 | |||
S75169.1 | - | 3 | 323 | |||
U01025.1 | - | 3 | 323 | |||
X62741.1 | - | 4 | 359 |