Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006868.3 | NP_006859.2 | 6 | 579 | ||
XM_017025528.1 | XP_016881017.1 | 7 | 961 | |||
XM_017025529.1 | XP_016881018.1 | 7 | 949 | |||
GenBank mRNA | AF183421.1 | - | 6 | 532 | ||
AF234995.1 | - | 6 | 578 | |||
AF498957.1 | - | 6 | 401 | |||
AK223505.1 | - | 6 | 443 | |||
AK314809.1 | - | 6 | 537 | |||
BC001148.1 | - | 6 | 501 | 3534853 | ||
BT020027.1 | - | 6 | 401 | |||
BT020028.1 | - | 6 | 401 | |||
BU944638.1 | - | 6 | 645 | |||
CR407659.1 | - | 6 | 401 | |||
U57091.1 | - | 6 | 446 | |||
U59877.1 | - | 6 | 461 |