Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307930.1 | NP_001294859.1 | 4 | 1778 | ||
NM_016563.3 | NP_057647.1 | 5 | 1835 | |||
XM_011521660.2 | XP_011519962.1 | 6 | 1648 | |||
GenBank mRNA | AF233588.1 | - | 5 | 1672 | ||
BC053734.1 | - | 5 | 1683 | 5588236 |