Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017811.3 | NP_060281.2 | 5 | 1139 | ||
XM_005251496.2 | XP_005251553.1 | 4 | 1072 | |||
XM_011517949.1 | XP_011516251.1 | 4 | 1024 | |||
XM_017014860.1 | XP_016870349.1 | 3 | 937 | |||
GenBank mRNA | AJ240087.1 | - | 5 | 570 | ||
AK000426.1 | - | 5 | 756 | |||
AK225880.1 | - | 5 | 744 | |||
BC004862.2 | - | 5 | 746 | 3838157 | ||
BC047584.1 | - | 5 | 995 | 4827644 | ||
CR457233.1 | - | 5 | 565 |