Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018058.6 | NP_060528.3 | 15 | 2719 | ||
GenBank mRNA | AJ276171.1 | - | 15 | 2512 | ||
AJ279016.1 | - | 14 | 2339 | |||
AK001182.1 | - | 12 | 2008 | |||
AK057190.1 | - | 15 | 2719 | |||
AK292084.1 | - | 15 | 2481 | |||
BC034245.1 | - | 13 | 1981 | 4178363 |