Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048241.2 | NP_001041706.1 | 4 | 270 | ||
NM_024292.3 | NP_077268.1 | 4 | 338 | |||
GenBank mRNA | AF313915.1 | - | 4 | 219 | ||
AK128885.1 | - | 4 | 338 | |||
AK309258.1 | - | 3 | 462 | |||
AK311889.1 | - | 4 | 258 | |||
AV682189.1 | - | 4 | 270 | |||
BC007053.1 | - | 4 | 292 | 3827384 | ||
BC110839.1 | - | 4 | 186 | 6474495 | ||
BP213482.1 | - | 4 | 258 | |||
BT007355.1 | - | 3 | 154 |