Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031885.3 | NP_114091.3 | 1, 2 | 346 | ||
XM_005256080.2 | XP_005256137.1 | 1, 2 | 345 | |||
GenBank mRNA | AF342736.1 | - | 1, 2 | 533 | ||
AK027635.1 | - | 1, 2 | 274 | |||
AK289604.1 | - | 1, 2 | 236 | |||
AK309068.1 | - | 1, 2 | 246 | |||
BC014140.2 | - | 1, 2 | 236 | 4653315 | ||
DA437930.1 | - | 1, 2 | 349 |