Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031885.3 | NP_114091.3 | 5 | 794 | ||
XM_005256080.2 | XP_005256137.1 | 5 | 793 | |||
GenBank mRNA | AB208905.1 | - | 5 | 574 | ||
AF342736.1 | - | 5 | 981 | |||
AK027635.1 | - | 5 | 722 | |||
AK289604.1 | - | 5 | 684 | |||
AK309068.1 | - | 5 | 694 | |||
BC014140.2 | - | 5 | 684 | 4653315 |