Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153712.4 | NP_714923.1 | 7 | 3663 | ||
XM_005263599.3 | XP_005263656.1 | 8 | 3822 | |||
XM_011510665.2 | XP_011508967.1 | 6 | 3558 | |||
GenBank mRNA | AB071393.2 | - | 8 | 3182 | ||
AK026673.1 | - | 1 | 1712 | |||
AK126378.1 | - | 2 | 2882 | |||
AK126549.1 | - | 5 | 3155 | |||
BX538316.1 | - | 3 | 2875 | |||
BX647135.1 | - | 4 | 2961 | |||
BX649144.1 | - | 1 | 8039 |