Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005614.3 | NP_005605.1 | 8 | 955 | ||
XM_011516457.1 | XP_011514759.1 | 9 | 1009 | |||
GenBank mRNA | AF148645.1 | - | 8 | 697 | ||
AF493921.1 | - | 8 | 542 | |||
AK125446.1 | - | 8 | 953 | |||
BC016155.1 | - | 8 | 955 | 3528583 | ||
BC066307.1 | - | 1 | 713 | 5274556 | ||
BC107705.1 | - | 1 | 904 | 6615350 | ||
BT006958.1 | - | 8 | 542 | |||
D78132.1 | - | 8 | 559 | |||
Z29677.1 | - | 8 | 565 |