Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015983.3 | NP_057067.1 | 6 | 469 | ||
XM_006715734.3 | XP_006715797.1 | 6 | 469 | |||
XM_011515420.2 | XP_011513722.1 | 7 | 562 | |||
XM_011515422.2 | XP_011513724.1 | 5 | 406 | |||
XM_011515423.2 | XP_011513725.1 | 3 | 253 | |||
XM_017012292.1 | XP_016867781.1 | 7 | 562 | |||
XM_017012293.1 | XP_016867782.1 | 7 | 562 | |||
XM_017012294.1 | XP_016867783.1 | 7 | 562 | |||
XM_017012295.1 | XP_016867784.1 | 4 | 316 | |||
GenBank mRNA | AF125044.1 | - | 6 | 508 | ||
AF125045.1 | - | 5 | 355 | |||
AK001446.1 | - | 6 | 457 | |||
BC004104.2 | - | 6 | 461 | 3835609 | ||
BG499454.1 | - | 6 | 484 |