Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004542.3 | NP_004533.1 | 4 | 322 | ||
XM_017026833.1 | XP_016882322.1 | 5 | 739 | |||
GenBank mRNA | AF044955.1 | - | 4 | 314 | ||
AF070653.1 | - | 4 | 323 | |||
AK289489.1 | - | 4 | 320 | |||
BC022369.1 | - | 4 | 319 | 4737656 | ||
BC061644.1 | - | 4 | 322 | 5748578 |